Silberhexafluoroantimonat ≥98%
Lieferant: Strem Chemicals, Inc.
|
Gefahr
|
Synonyme:
Silver hexafluoroantimonate(V)
93-4748-1G
STRE93-4748-1GEA
39.2
EUR
STRE93-4748-1G
Silberhexafluoroantimonat ≥98%
Silberhexafluoroantimonat
Formel:
AgF₆Sb Molecular Weight: 343.62 Dichte: 4.65…4.85 g/cm³ (20 °C) |
MDL:
MFCD00003401 CAS-Nummer: 26042-64-8 EINECS: 247-429-1 UN: 3260 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...