Tetrapropylzinn ≥97%

Lieferant: Strem Chemicals, Inc.
Achtung

93-5053-1G
STRE93-5053-1GEA 51.5 EUR
STRE93-5053-1G
Tetrapropylzinn ≥97%
Tetrapropylzinn
Formel: C₁₂H₂₈Sn
Molecular Weight: 291.06 g/mol
Siedepunkt: 222 °C
Dichte: 1.1065 (20°)
MDL: MFCD00015196
CAS-Nummer: 2176-98-9
EINECS: 218-536-0
UN: 2788
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR