Gadoliniumtrinitrat Hexahydrat ≥99,9% (Gd-Basis)
Lieferant: Strem Chemicals, Inc.
|
Gefahr
|
Synonyme:
Gadolinium (III) nitrate hexahydrate
93-6417.0100
STRE93-6417.0100EA
265
EUR
STRE93-6417.0100
Gadoliniumtrinitrat Hexahydrat ≥99,9% (Gd-Basis)
Gadoliniumtrinitrat Hexahydrat
Formel:
GdN₃O₉·6H₂O Molecular Weight: 451,36 g/mol Schmelzpunkt: 91,5…92 °C Dichte: 2,4 g/cm³ (20 °C) |
MDL:
MFCD00149728 CAS-Nummer: 19598-90-4 EINECS: 233-437-2 UN: 1477 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...