Rat recombinant biliverdin reductase
Lieferant: Enzo Life Sciences (Stressgen)
OSP-400B
STSSOSP-400BEA
0
EUR
STSSOSP-400B
Rat recombinant biliverdin reductase
Enzyme
Biliverdin reductase (phosphoprotein BVR) is an oxireductase which catalyses the reduction of the gamma-meso bridge of biliverdin to bilirubin.