Bovine native alpha-A-Crystallin
Lieferant: Enzo Life Sciences (Stressgen)
SPP-226F
STSSSPP-226FEA 0 EUR
STSSSPP-226F
Bovine native alpha-A-Crystallin
Proteine und Peptide
Alpha A crystallins (known as CRYAA, HspB4, heat shock protein β-4) are major water-soluble proteins accounting for almost 50% of total protein in the mammalian transparent eye lens and they are also found in a variety of other tissues.
  • Isolated from bovine eye lens
  • Purified by multi-step chromatography (SDS-PAGE, western blot)
  • Recommended dilutions (conditions) include western blot (100 ng/lane, colorimetric)
It has a liquid formulation in PBS at pH 7,2 containing 0,15M sodium chloride and 0,05M phosphate buffer.