Bovine native alpha-B-Crystallin
Lieferant: Enzo Life Sciences (Stressgen)
SPP-227F
STSSSPP-227FEA 0 EUR
STSSSPP-227F
Bovine native alpha-B-Crystallin
Proteine und Peptide
Alpha B crystallins (known as CRYAB, HspB5, Heat shock protein β-5) are major water-soluble proteins accounting for almost 50% of total protein in the mammalian transparent eye lens and they are also found in a variety of other tissues.
  • Isolated from bovine eye lens
  • Purified by multi-step chromatography (SDS-PAGE, western blot)
  • Recommended dilutions (conditions) include western blot (50 ng/lane, colorimetric)
It has a liquid formulation in PBS at pH 7,2 containing 0,09% sodium azide.