Bovine Recombinant HSC70/HSP73 (from E. coli)
Lieferant: Enzo Life Sciences (Stressgen)
SPP-751D
STSSSPP-751DEA 0 EUR
STSSSPP-751D
Bovine Recombinant HSC70/HSP73 (from E. coli)
Proteine und Peptide
The Hsp70 family of heat shock proteins contains multiple homologs ranging in size from 66-78 kDa, and are the eukaryotic equivalents of the bacterial DnaK. Hsp70 nucleotide exchange and substrate binding coordinates the folding of newly synthesised proteins, the re-folding of misfolded or denatured proteins, coordinates trafficking of proteins across cellular membranes, inhibits protein aggregation, and targets the degradation of proteins via the proteasomal pathway.
  • Produced in E. coli
  • Purified by multi-step chromatography (SDS-PAGE, western blot)
It has a liquid formulation in 30 mM TRIS-HCl at pH 7,5, containing 1,0 mM EDTA, 2,0 mM DTT, and 300 mM sodium chloride.