Bovine Recombinant HSC70/HSP73 (from E. coli), Biotin
Lieferant: Enzo Life Sciences (Stressgen)
The Hsp70 family of heat shock proteins contains multiple homologs ranging in size from 66 to 78 kDa, and are the eukaryotic equivalents of the bacterial DnaK. Hsp70 nucleotide exchange and substrate binding coordinates the folding of newly synthesised proteins, the re-folding of misfolded or denatured proteins, coordinates trafficking of proteins across cellular membranes, inhibits protein aggregation, and targets the degradation of proteins via the proteasomal pathway.
- Produced in E. coli
- Purified by multi-step chromatography (SDS-PAGE, western blot)
It has a liquid formulation in 10 mM sodium phosphate at pH 7,2.