Cole-Parmer® Stuart V Vortex Mixers
Lieferant: Cole Parmer
04729-03 04729-01 04729-07 04729-13
STUA04729-03EA 606 EUR
STUA04729-03 STUA04729-01 STUA04729-07 STUA04729-13
Cole-Parmer® Stuart V Vortex Mixers
Schüttler Vortex-Schüttler
Compact unit perfect for individual work stations.
  • Mix a wide variety of tube sizes and shapes with the included rubber cup head
  • Heavy die-cast body won't dance across the bench
  • Automatic start when the soft rubber cup is pressed down
The vortex mixer provides rapid mixing of samples contained in test-tubes, small flasks, and bottles. Starts automatically when the rubber cup is depressed and stops once vessel is removed. Heavy die-cast body for stability during use.
The fixed speed Stuart Vortex Mixer measures 7.8×7.8×21.5 cm (W×H×D) with a 4.2 mmMore Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable
Max. load 1 kg
Kapazität 1 tube