Cole-Parmer® ST-200 Series Undergrad Analogue Magnetic Stirrer
Lieferant: Cole Parmer
04807-56
STUA04807-56EA 472 EUR
STUA04807-56
Cole-Parmer® ST-200 Series Undergrad Analogue Magnetic Stirrer
Rührer Magnetrührer
Powerful and precision stirring in compact size.
  • Built with tough aluminium casing for longevity
  • Designed to prevent accidental spillage on control panel
  • IP 32 rated for water and dust resistance
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
Constructed with powerful magnets and motor, these analogue stirrers provide stirring speeds of up to 2000 min⁻¹ and mixing capability of up to 15 L of liquid. The compact, stackable design takes up less benchtop and storage space.
For safety, all units are designed to direct spills away from the user controls. The stirrers also feature BioCote® antimicrobial protection which reduces the presence of odour and stain causing microbes such as bacteria, mould, and fungi on the product surface by up to 99,99%.