Cole-Parmer® ST-200 Series Large-Capacity Magnetic Stirrers
Lieferant: Cole Parmer
04807-74
STUA04807-74EA 1550 EUR
STUA04807-74
Cole-Parmer® ST-200 Series Large-Capacity Magnetic Stirrers
Rührer Magnetrührer
Powerful magnets for strong magnetic coupling.
  • Large plate area accommodates one large vessel up to 30 L
  • Easy-to-use stirring control dial
  • Robust, durable construction with stainless steel top
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
These general-purpose stirrers are designed with safety as well as performance in mind. Powerful magnets and motor give stirring speeds up to 600 min⁻¹ and can mix up to 30 L easily. The electronic feedback speed control accurately maintains the set speed.
Units feature BioCote antimicrobial protection which reduces the presence of odour and stain causing microbes such as bacteria, mould, and fungi on the product surface by up to 99.99%.