Cole-Parmer® WB-300 Series General-Purpose Digital Water Baths, Stuart
Lieferant: Cole Parmer
12122-91 12122-71 12122-81
STUA12122-91EA 1240 EUR
STUA12122-91 STUA12122-71 STUA12122-81
Cole-Parmer® WB-300 Series General-Purpose Digital Water Baths, Stuart
Bäder Wasserbäder, ohne Umwälzung
Easy to use and easy to clean.
  • Analogue dial with digital temperature display makes it simple to use
  • Safety features include low water level sensor and over temperature protection
  • Stainless steel tank with rounded corners will not trap dirt
  • Integral drain simplifies water changes and cleaning
These simple to use heated water baths feature an LED temperature display with an analogue dial for quick and easy set point adjustments. Low water level sensor prevents dry boiling, and over temperature protection system monitors temperature and controls heaterMore Product Information