Cole-Parmer® Digital Water Baths
Lieferant: Cole Parmer
16340-65
STUA16340-65EA 1290 EUR
STUA16340-65
Cole-Parmer® Digital Water Baths
Bäder Wasserbäder, ohne Umwälzung
Pair with Stuart rotary evaporators for maximum efficiency and smallest footprint.
  • Precise digital temperature display
  • Corrosion-resistant bowl
  • Easy to clean
  • Integrated carry handle for water bath
  • Cool-touch outer case
The clear LED digital display shows the actual bath temperature or the set temperature. There are also two water bath options. The 90 °C bath has a 304 stainless steel bowl and is for water only, suitable for low boiling point applications. The higher temperature 180 °C bath has an aluminium bowl with PTFE coating and can be used with both water and oil, ideal for higher boiling point liquids.
Bitte beachten Use only water bath model 16340-64 with plastic-coated glassware since it is limited to 80 °C.