Cole-Parmer® WA-200 Wrist-Action Shaker
Lieferant: Cole Parmer
51900-02
STUA51900-02EA 2740 EUR
STUA51900-02
Cole-Parmer® WA-200 Wrist-Action Shaker
Schüttler Rotations-/Wippschüttler
Eliminate wrist strain.
  • Vigorous mixing action simulates hand shaking
  • Ideal for extractions
  • Accepts up to 500 ml flasks
The variable speed shaker features two side arms with adjustable clamps capable of holding eight flasks or bottles up to 500 ml capacity. Use for prolonged continuous shaking or set the built-in analog timer from 10 to 60 minutes and unit automatically turns off when done. Electronic feedback controller ensures a constant speed, irrespective of load.
Lieferumfang: Includes two side arms, eight clamps, allen key, and power cord with European plug.