Cole-Parmer® SH-200 Gyro Rocking Shakers
Lieferant: Cole Parmer
51901-12 51901-04
STUA51901-12EA 1850 EUR
STUA51901-12 STUA51901-04
Cole-Parmer® SH-200 Gyro Rocking Shakers
Schüttler Rotations-/Wippschüttler
Gentle swirling prevents foaming.
  • Adjust tilt angle for optimal mixing
  • Platform lip and non-slip mat hold vessels securely in place
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be used in incubators
These gyro rocking shakers feature a 3D gyrating action with a 3 to 12° tilt angle. Easily adjust the angle of the tilt by hand for optimal performance. Best suited for low-foaming requirements, the shakers are ideal for cell culturing, staining, and destaining. Easily set and view the speed and the timer on the LED display. Control speed from 5 to 70 min⁻¹. The microprocessor maintains speed accuracy even over long runs. Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert.
Durable shakers are rated for continuous use. Use in incubators and environmental or cold chambers up to 40 °C.
Lieferumfang: Units include flat platform, non-slip mat, and power cord.