Mixer designed for reliability and durability.
- Ideal for light-duty mixing of water-like fluids
- Brushed DC motor with integrated overload protection
- Durable - no wearing parts that require user service
- User-replaceable keyless chuck
- 1/50 hp brushed DC motor
The Cole-Parmer GP laboratory mixer ensures accurate, dependable motor speed control with 'soft-start' feature and smooth, gradual adjustment to the speed setting. Constant speed is maintained electronically via automatic compensation for changes in load torque demands at both high and low speeds. Built-in motor current compensation minimises speed drift with changes in load. Motor current limiting protects the system in case of a locked rotor condition. Output circuit protection protects against inadvertent output shorting.
The mixer rotates at 40 to 4000 min⁻¹ with a maximum torque of 9,5 in-oz (6,7 N·cm). The speed control knob is used to turn on and set the speed of the mixer motor and provides a 100:1 adjustment range. The LED indicates the mixer is powered on when it is green. When it flashes, it indicates a limit condition has been reached, such as an over temperature or over-current situation.
Shaft: Lר: 305×10 mm
Motor: L×H 84×104 mm
Material: Painted or anodised aluminum, plastic, and vinyl
Standards: IP protection class according to DIN EN 60529: IP 44.
Lieferumfang: Includes Mixer motor with integrated controller and 183 cm (6') cable, 12× 0.95 cm (³/₈") stainless steel shaft, 380 mm (1,5") stainless steel propeller, mounting rod and power cable.