Cole-Parmer® Generator Probe Tool Kit for Homogenisers HO-200 and HO-400
Lieferant: Cole Parmer
78905-47
STUA78905-47EA 0 EUR
STUA78905-47
Cole-Parmer® Generator Probe Tool Kit for Homogenisers HO-200 and HO-400
Elektroden Elektroden, sonstige
Keep your homogeniser up and running.
  • Optimise performance of your homogeniser generator probes
Ideal replacement parts for Cole-Parmer branded homogenisers and probes.
Lieferumfang: Supplied with knife tool to remove 5, 7, and 10 mm probe knives, bearing removal tools for 5 and 7 mm generator probes and a shaft holder.