Aluminium-Standardlösung, 1.000 mg/l Al in verd. Salpetersäure (aus Al(NO3)3.9H2O) TraceCERT® Standard für die ICP, Supelco®
Lieferant: Merck
61935-100ML-F
SUPL61935-100ML-FEA
112
EUR
SUPL61935-100ML-F
Aluminium-Standardlösung, 1.000 mg/l Al in verd. Salpetersäure (aus Al(NO3)3.9H2O) TraceCERT® Standard für die ICP, Supelco®
Standards
Kationischer Standards
Aluminium-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the certificate.