Drug Test, 6 Parameter Multi-Panel Dip Urine
Lieferant: SURESCREEN
S6DS13
SURES6DS13EA 212 EUR
SURES6DS13
Drug Test, 6 Parameter Multi-Panel Dip Urine
Drogentests
AMP/BZO/BUP/COC/MTD/OPI six parameter multi-panel dip urine drug testdrug test with parameter.