Schneidwerkzeug
Lieferant: SWANN MORTON
1231
SWAN1231EA 68.7 EUR
SWAN1231
Schneidwerkzeug
Skalpelle Skalpelle, komplett
Klinge aus Stahl, mit Kunststoff-Handgriff. Das ideale Werkzeug für viele Schneid- und Modellierarbeiten.
  • Thumbscrew is used for blade changing and retaining
  • Handle made from PS is impact resistant
Three types of replacement blades available as an option
Bestellinformation: This starter set includes the plastic handle and two blades (one each No. 1 and No. 2 (carded)). Handle and blades are also available separately.