Surgical blades for cervical biopsy
Lieferant: SWANN MORTON
2001
SWAN2001EA 135 EUR
SWAN2001
Surgical blades for cervical biopsy
Skalpelle Skalpellklingen
This sterile stainless cervical biposy blade is used in conjunction with the long stainless steel Major handle to take a 'cone' biopsy from the tissue in a woman's cervix. The tissue sample can then be analysed to assess the prescence of malignant and non malignant carcinoma cells.
  • Fits handles Major S and L