Surgical scalpel handle, SF13
Lieferant: SWANN MORTON
6061
SWAN6061EA 76.3 EUR
SWAN6061
Surgical scalpel handle, SF13
Skalpelle Skalpellgriffe
Stainless steel.
Fits blades SM61, SM62, SM63, SM64, SM65, SM65A, SM67, SM68, SM69, SP90, SP91 and Myringotomy blade.
Verpackung: Individually sleeved.