Thymol ≥98,5%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Thymol is generally added in the Hematoxylin and Eosin staining solution to prevent mould growth. It is also suitable for use in antimicrobial screening by the disk diffusion method. It has been used as a test compound in antifungal assay to assess the susceptibility of various fungal isolates.
Formel:
2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH Siedepunkt: 232 °C (Lit.) Schmelzpunkt: 48…51 °C (Lit.) Dichte: 0,965 g/cm³ (25 °C) (Lit.) Flammpunkt: 116 °C (geschlossener Tiegel) Lagertemperatur: Room Temperature |
MDL:
MFCD00002309 CAS-Nummer: 89-83-8 EINECS: 201-944-8 UN: 2430 ADR: 8,III Merck Index: 13,09476 |
|
Spezifikation Testergebnisse
Appearance (Colour) | Colorless to White |
Appearance (Form) | Powder or Crystals |
Solubility (Colour) | Colorless |
Solubility (Turbidity) 50 mg/ml, EtOH |
Clear |
1H NMR Spectrum | Conforms to Structure |
Purity (HPLC) | ≥98.5 % |
Recommended Retest Period | 3 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...