Tetramethylrhodamine-5-isothiocyanate (5-TRITC), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Xanthylium
M-Clarity™ Qualitätsstufe = MQ200
T0820-5MG
T0820-5MGEA
360
EUR
T0820-5MG
Tetramethylrhodamine-5-isothiocyanate (5-TRITC), Sigma-Aldrich®
Tetramethylrhodamine-5-isothiocyanate (5-TRITC)
Formel:
C₂₅H₂₁N₃O₃S Molecular Weight: 443,52 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD07438035 CAS-Nummer: 80724-19-2 |
Spezifikation Testergebnisse
Appearance (Color) | Pink to Purple |
Appearance (Form) | Powder |
Proton NMR spectrum | Conforms to Structure |
EMM (in MeOH) | 82 - 98 (540 - 546 nm) |
Fluorescence (Emission max) | 567 - 575 nm |
Purity (HPLC) | ≥ 97% at 254 nm (Isomer purity) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...