Thioflavin S, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: C.I. 49010, Direct Yellow 7

T1892-25G
T1892-25GEA 83.8 EUR
T1892-25G
Thioflavin S, Sigma-Aldrich®
Thioflavin S

Thioflavine S has been used to stain and visualize β-amyloid structures in the tissue sections.

Lagertemperatur: Raumtemperatur
MDL: MFCD00071453
CAS-Nummer: 1326-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR