Thymol iodide, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

T2763-25G
T2763-25GEA 240 EUR
T2763-25G
Thymol iodide, Sigma-Aldrich®
Thymol iodide

Thymol iodide, (4,4'-Bis(iodooxy)-2,2'-dimethyl-5,5'-bis(1-methylethyl)-1,1'-biphenyl)

Formel: C₂₀H₂₄I₂O₂
Molecular Weight: 550,219 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00026409
CAS-Nummer: 552-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1feecd85d1195da8aa3a9c4035747c882d242dae&vid=932940a53fdeaf3c2103e8cbf17de620b533f2a3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR