N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid ≥95%, Pulver, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ200

T3134-5G
T3134-5GEA 78.3 EUR
T3134-5G
N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid ≥95%, Pulver, Sigma-Aldrich®
N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid
Formel: C₁₀H₁₈Cl₂N₂
Schmelzpunkt: 222,..224 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00012482
CAS-Nummer: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdb59462690da7f44f0ba4e2ee465bb1fa318e61&vid=f952549e50e1861aa87fb1b41ad0a9689a7cd94e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Off-White to Grey to Brown
Appearance (Form) Powder
Infrared Spectrum Conforms to Structure
Purity (HPLC) ≥ 95 %
Titration with HClO4 ≥ 99 %
Solubility (Colour) Colorless to Light Purple to Dark Brown
Solubility (Turbidity)
5 g/100 mL, H2O
Clear to Very Slightly Hazy
Recommended Retest Period 2 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR