Trenbolon, Supelco®

Lieferant: Merck

T3925-250MG
T3925-250MGEA 181 EUR
T3925-250MG
Trenbolon, Supelco®
Trenbolon
Formel: C₁₈H₂₂O₂
MDL: MFCD00214395
CAS-Nummer: 10161-33-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=965a3e1bb952689e40931c3466a3f643217e6aea&vid=53c2392d2eb1e09ec6d45d99e608dde06f9c8638&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR