JC-1 (iodide), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: JC1, 5,5',6,6'-Tetrachloro-1,1',3,3'-tetraethylbenzimidazolocarbocyanine iodide, 1,1',3,3'-tetraethyl-5,5',6,6'-tetrachloroimidacarbocyanine iodide, 5,5’,6,6’-Tetrachloro-1,1’,3,3’-tetraethylbenzimidazolylcarbocyanine iodide

T4069-5MG
T4069-5MGEA 870 EUR
T4069-5MG
JC-1 (iodide), Sigma-Aldrich®
JC-1 (iodide)

A dual-emission potential-sensitive probe that can be used to measure mitochondrial membrane potential. JC-1 is a green-fluorescent (λex 520 nm) monomer at low membrane potential. At higher potentials, JC-1 forms red-fluorescent (λem 596 nm ) "J-aggregates," which exhibit broad excitation and very narrow emission spectra. The ratio of red to green fluorescence of JC-1 is dependent only on membrane potential, and not influenced by mitochondrial size, shape, or density.

Formel: C₂₅H₂₇Cl₄IN₄
Molecular Weight: 652.223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00188475
CAS-Nummer: 3520-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c9e0dd188fb2aa7aed32d95bfdf6b98751e5b58&vid=03effc2ed578bbc962498627b52aeafc786f290c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR