Genuine Tecan® 1000 µL disposable automation tips for Freedom EVO® Liquid Handling Arm™ (LiHa) or Fluent® Flexible Channel Arm™ (FC A). Hanging tip format, non-filtered, conductive. Tecan PURE purity level - tested and certified to be free of hum Dn 1 * 2.304 ST
Lieferant: Tecan
Genuine Tecan® 1000 µL disposable automation tips for Freedom EVO® Liquid Handling Arm™ (LiHa) or Fluent® Flexible Channel Arm™ (FC A). Hanging tip format, non-filtered, conductive. Tecan PURE purity level - tested and certified to be free of hum Dn 1 * 2.304 ST