Genuine Tecan® 50 µL disposable automation tips designed specifically for use on the Freedom EVO® and Fluent® MultiChannel Arm™ (MC A) 96 option. ANSI format, single stack, non-filtered. Tecan PURE purity level - tested and certified to be free o as 1 * 3.840 ST
Lieferant: Tecan
Genuine Tecan® 50 µL disposable automation tips designed specifically for use on the Freedom EVO® and Fluent® MultiChannel Arm™ (MC A) 96 option. ANSI format, single stack, non-filtered. Tecan PURE purity level - tested and certified to be free o as 1 * 3.840 ST