Genuine Tecan® 100 µL disposable automation tips designed specifically for use on the Freedom EVO® and Fluent® MultiChannel Arm™ (M CA) 96 option. ANSI format, single stack, filtered. Tecan STERILE purity level - sterile, tested and certified to man 1 * 3.840 ST
Lieferant: Tecan
Genuine Tecan® 100 µL disposable automation tips designed specifically for use on the Freedom EVO® and Fluent® MultiChannel Arm™ (M CA) 96 option. ANSI format, single stack, filtered. Tecan STERILE purity level - sterile, tested and certified to man 1 * 3.840 ST