Genuine Tecan® 200 µL disposable automation tips for Freedom EVO® Liquid Handling Arm™ (LiHa) or Fluent® Flexible Channel Arm™ (FCA ). ANSI format, non-filtered, conductive. Tecan STERILE purity level - Sterile, tested and certified to be free fro a 1 * 1 SET
Lieferant: Tecan
Genuine Tecan® 200 µL disposable automation tips for Freedom EVO® Liquid Handling Arm™ (LiHa) or Fluent® Flexible Channel Arm™ (FCA ). ANSI format, non-filtered, conductive. Tecan STERILE purity level - Sterile, tested and certified to be free fro a 1 * 1 SET