Genuine Tecan® 1000 µL disposable automation tips for Freedom EVO® Liquid Handling Arm™ (LiHa) or Fluent® Flexible Channel Arm™ (FC A). ANSI format, non-filtered, conductive. Tecan STERILE purity level - Sterile, tested and certified to be free fr n 1 * 3.840 ST
Lieferant: Tecan
Genuine Tecan® 1000 µL disposable automation tips for Freedom EVO® Liquid Handling Arm™ (LiHa) or Fluent® Flexible Channel Arm™ (FC A). ANSI format, non-filtered, conductive. Tecan STERILE purity level - Sterile, tested and certified to be free fr n 1 * 3.840 ST