Cerex® Atlas® SPE Columns
Lieferant: Tecan
30163871 30163862 30163032 30160523 30165975 30165979 30168411 30168415 30203864 30168419 30176001 30168423
TCAT30163871EA 0 EUR
TCAT30163871 TCAT30163862 TCAT30163032 TCAT30160523 TCAT30165975 TCAT30165979 TCAT30168411 TCAT30168415 TCAT30203864 TCAT30168419 TCAT30176001 TCAT30168423
Cerex® Atlas® SPE Columns
Chromatographiesäulen
A hydrophilic/Lipophilic balanced chemistry offering selectivity to a wide range of analytes. It provides enhanced selectivity for polar species, also comes in larger particle for more viscous samples. A direct alternative to Waters Oasis® HLB and Phenomenex Strata X.
  • Mode: Reversed-phased
Atlas® a hydrophilic-lipophilic balanced polymeric sorbent is a N-vinylpyrrolidone-divinylbenzene co-polymer. It provides selectivity for both polar and non-polar analytes thus allowing the user to extract a variety of samples with ease. The divinylbenzeneMore Product Information