Atlas® NBE SCX SPE Plates, 96-well
Lieferant: Tecan
30174724 30174732
TCAT30174724EA 0 EUR
TCAT30174724 TCAT30174732
Atlas® NBE SCX SPE Plates, 96-well
Mikrotiterplatten Festphasenextraktionsplatten
A hydrophilic/lipophilic balanced strong cationic chemistry offering excellent selectivity to basic and neutral compounds. Also utilising Tecan's unique NBE column platform. A direct alternative to Waters Oasis® MCX and Phenomenex Strata XC.
  • Base resin chemistry: N-vinylpyrrolidonedivinylbenzene copolymer
  • Surface chemistry: sulfonic acid
  • Particle packing: monodisperse porous microsphere
  • pH stability: 1 - 14
  • Simplified sample prep. protocol
  • Slurry packing ensures highest of quality, consistency and reproducibility
  • Available with SCX chemistry
  • NBE format provides greater extraction efficiency
Atlas® SCX incorporates this copolymer technology along with a strong cationic exchange chemistry to provide a focused clean-up when working with basic compounds in tough matrices. In addition to the balanced copolymer technology, Atlas® SCX provides an additional chemistry that allows for strong retention of basic compounds.
Ideal for use in basic small molecules and pharmaceutical drugs, sample preparation for drug monitoring, screening, peptide clean-up, etc., biopharma/life science and food safety.