Cerex® FLORASIL SPE Columns
Lieferant: Tecan
30213512
TCAT30213512EA 0 EUR
TCAT30213512
Cerex® FLORASIL SPE Columns
Chromatographiesäulen
Florisil is used for sample preparation (solid phase extraction) of e.g., organic tin compounds and aliphatic carboxylic acids. Florisil is a magnesium silicate.
  • Ion-exchange
  • Liquid handler compatible