SPE Column, NBE HPSAX
Lieferant: Tecan
412-0021R-NBE
TCAT412-0021R-NBEEA 0 EUR
TCAT412-0021R-NBE
SPE Column, NBE HPSAX
Chromatographiesäulen
Polymeric strong anionic exchange chemistry.
  • Ion-exchange
  • Liquid handler compatible
20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.