SPE Plate, 96-well, NBE HPSAX
Lieferant: Tecan
412-0051NBE
TCAT412-0051NBEEA 0 EUR
TCAT412-0051NBE
SPE Plate, 96-well, NBE HPSAX
Mikrotiterplatten Festphasenextraktionsplatten
Polymeric strong anionic exchange chemistry.
  • Ion-exchange
  • Liquid handler compatible
20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.