Cerex® HPSAX SPE Columns
Lieferant: Tecan
412-0101RCH 412-0101R 412-0201R 412-0401R 412-0353C
TCAT412-0101RCHEA 0 EUR
TCAT412-0101RCH TCAT412-0101R TCAT412-0201R TCAT412-0401R TCAT412-0353C
Cerex® HPSAX SPE Columns
Chromatographiesäulen
Polymeric strong anionic exchange chemistry.
  • Ion-exchange
  • Liquid handler compatible
20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.