Cerex® WWP2 96 Well Plate, RCH
Lieferant: Tecan
417-0101RCH
TCAT417-0101RCHEA 0 EUR
TCAT417-0101RCH
Cerex® WWP2 96 Well Plate, RCH
Mikrotiterplatten Festphasenextraktionsplatten
Polymeric water-wettable resin ideal for reverse-phase retention of aqueous samples.
  • Reversed-phase
  • Liquid handler compatible
Special water wettable polymer with 300 Å pore size ideal for protein digest clean up and other protein related workflows.
Verpackung: 1 piece per box