Cerex® HPSCX SPE Columns
Lieferant: Tecan
427-0353C 427-0506C
TCAT427-0353CEA 0 EUR
TCAT427-0353C TCAT427-0506C
Cerex® HPSCX SPE Columns
Chromatographiesäulen
Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.
  • Ion-exchange
  • Liquid handler compatible
Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable