96 Well Plate, RCH TRACE-N20
Lieferant: Tecan
611-0101RCH
TCAT611-0101RCHEA 0 EUR
TCAT611-0101RCH
96 Well Plate, RCH TRACE-N20
Mikrotiterplatten Festphasenextraktionsplatten
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
  • Reversed-phase
  • Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.