SPE Plates, 96-well, NBE MAESTRO
Lieferant: Tecan
613-0051NBE
TCAT613-0051NBEEA 0 EUR
TCAT613-0051NBE
SPE Plates, 96-well, NBE MAESTRO
Mikrotiterplatten Festphasenextraktionsplatten
Silica-based sorbent with C18 bonding in proprietary coverage.
  • Reversed-phased
  • Liquid handler compatible
Uses a combination of filtration and sorbent interaction. Ideal for pass-through sample preparation techniques to remove phospholipids, proteins and particulates from sample matrices.