SPE Plate, 96-well, NBE OFX
Lieferant: Tecan
633-0031NBE
TCAT633-0031NBEEA 0 EUR
TCAT633-0031NBE
SPE Plate, 96-well, NBE OFX
Mikrotiterplatten Festphasenextraktionsplatten
A proprietary polymer resin blend of ion-exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.