SPE Plate, 96-well, NBE OFQC
Lieferant: Tecan
637-0051NBE
TCAT637-0051NBEEA 0 EUR
TCAT637-0051NBE
SPE Plate, 96-well, NBE OFQC
Mikrotiterplatten Festphasenextraktionsplatten
A proprietary polymer resin blend of ion-exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.