Cerex® ETGS SPE Column
Lieferant: Tecan
642-0501R
TCAT642-0501REA 0 EUR
TCAT642-0501R
Cerex® ETGS SPE Column
Chromatographiesäulen
Specialized silica-based anion exchange chemistry designed for ETG/ETS testing in biofluids.
  • Ion-exchange
  • Liquid handler compatible
A 10 µm micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for ETS/ETGS Testings.