96 Well Plate, RCH CEREX FNT
Lieferant: Tecan
667-0101RCH
TCAT667-0101RCHEA 0 EUR
TCAT667-0101RCH
96 Well Plate, RCH CEREX FNT
Mikrotiterplatten Festphasenextraktionsplatten
A 15 µm micro particulate with strong cation exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
  • Cerex
  • Ion-exchange
  • Liquid handler compatible
Formulated for FNT and pain panel.
Verpackung: 1 piece per box