SPE Plate, 96-well, NBE PWCX
Lieferant: Tecan
675-0051NBE
TCAT675-0051NBEEA 0 EUR
TCAT675-0051NBE
SPE Plate, 96-well, NBE PWCX
Mikrotiterplatten Festphasenextraktionsplatten
Polymeric, weak cation exchange chemistry for strong bases.
  • Ion-exchange
  • Liquid handler compatible
A 19 µm polymer micro particulate with weak cationic exchange chemistry providing large contac zone for sorbent analyte interaction resulting in a fast and uniform sample prep.