96 Well Plate, RCH PWCX
Lieferant: Tecan
675-0201RCH
TCAT675-0201RCHEA 0 EUR
TCAT675-0201RCH
96 Well Plate, RCH PWCX
Mikrotiterplatten Festphasenextraktionsplatten
Polymeric, weak cation exchange chemistry for strong bases.
  • Ion-exchange
  • Liquid handler compatible
A 19 µm polymer micro particulate with weak cationic exchange chemistry providing large contac zone for sorbent analyte interaction resulting in a fast and uniform sample prep.